Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Polar Fused Silica Tubing

$54.50 - $386.00

Specifications

Type Tubing
Material Fused Silica
For Use With (Application) Gas Chromatography (GC)
Description Composition: PEG (Deactivated)
View More Specs

Products 10
Catalog Number Mfr. No. Diameter (Metric) Inner Overall Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner Overall Length (Metric) Price Quantity & Availability  
11-100-4755
View Documents
MilliporeSigma™ Supelco™
25710
0.10 mm 1 m Each for $71.50
Add to cart
 
11-100-4964
View Documents
MilliporeSigma™ Supelco™
25712
0.25 mm 1 m Each for $54.50
Add to cart
 
11-100-3938
View Documents
MilliporeSigma™ Supelco™
25777
0.25 mm 30 m Each for $268.00
Add to cart
 
11-100-4819
View Documents
MilliporeSigma™ Supelco™
25752U
0.32 mm 5 m Each for $65.90
Add to cart
 
11-100-4306
View Documents
MilliporeSigma™ Supelco™
25765
0.32 mm 15 m Each for $159.00
Add to cart
 
11-100-4679
View Documents
MilliporeSigma™ Supelco™
25714
0.53 mm 1 m Each for $79.50
Add to cart
 
11-100-4833
View Documents
MilliporeSigma™ Supelco™
25734
0.53 mm 3 m Each for $64.30
Add to cart
 
11-100-4675
View Documents
MilliporeSigma™ Supelco™
25753
0.53 mm 5 m Each for $79.50
Add to cart
 
11-100-4146
View Documents
MilliporeSigma™ Supelco™
25766
0.53 mm 15 m Each for $204.00
Add to cart
 
11-100-3752
View Documents
MilliporeSigma™ Supelco™
25779
0.53 mm 30 m Each for $386.00
Add to cart
 
Specifications

Specifications

Tubing
Fused Silica
Gas Chromatography (GC)
Composition: PEG (Deactivated)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit