Learn More
Thermo Scientific™ Polara™ RS Software
Transform any RapidStak™ microplate stacker into a complete, optimized benchtop assay solution with Thermo Scientific™ Polara RS Software.
Manufacturer:  Thermo Scientific™ F01543
Catalog No. 14-386-553
Part Not Found
Description
Description
Transform any RapidStak™ microplate stacker into a complete, optimized benchtop assay solution with Thermo Scientific™ Polara RS Software. Polara RS allows you to program your RapidStak to perform almost any plate-loading application, from bulk multi-reagent dispensing to shoot and read assays to ELISA preparation. Polara RS allows you to automate an endless variety of one or two-instrument assays.
  • Full, intuitive instrument control, with drag and drop GUI
  • Can control, automate, and schedule most lab instruments
  • Compatible with the majority of microplate instruments and plates
  • Enables use of the RapidStak for timed incubation steps in the assay
  • Easy field installation or upgrade
  • Error handling/recovery and barcode reading
  • Offline simulations to maximize reagent and instrument usage
  • Any RapidStak can run Polara RS, no hardware or factory upgrade required
Works with the Following Thermo Scientific Instruments:
  • Multidrop Combi
  • Multidro Combi nL
  • Multidrop 384
  • Wellwash Versa
  • Multiskan GO
  • Multiskan FC
  • Varioskan Flash
  • SkanIt readers
  • Ascent readers
  • ALPS 3000
  • ALPS 300
  • Seal-It 100
Specifications
Specifications
Multidrop Combi; Multidro Combi nL; Multidrop 384; Wellwash Versa; Multiskan Go; Multiskan FC; Varioskan Flash; Skanit Readers; Ascent Readers; Alps 3000; Alps 300; Seal-It 100
Polara RS; Single to Dual Instrument License