Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Argos Technologies PolarSafe™ 81-Place Polypropylene Storage Boxes

Used for freezer storage and organization. Argos Technologies PolarSafe™ 81-Place Polypropylene Storage Boxes holds 0.5, 1.5, and 2.0mL microtubes as well as most brands of vials.

Manufacturer:  Argos Technologies R3120

 View more versions of this product

Catalog No. 03-391-549


  Check Availability
Add to cart

Description

Description

  • Durable storage boxes for freezer storage and organization
  • Matte surface on box sides for printing and or labeling
  • Withstands temperatures from -80°C to +121°C
  • Alphanumeric grid for sample identification
  • Fits most standard 2 in. stainless steel freezer racks
Specifications

Specifications

Orange
Storage box
0.5, 1.5, and 2.0mL tubes
5
Polypropylene
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit