Learn More
Cole-Parmer™ PolarSafe™ Cooling Packs and Accessories
Transport your critical samples without worry
 View more versions of this product
Catalog No. 13-200-152
Add to cart
Description
Description
  • Use the cooling end-caps and cooling frames together to encapsulate the entire sample.
  • This modular system can accommodate various sample sizes to provide a complete thermal barrier.
  • Temperature ranges are 2 to 8°C and 20 to 24°C respectively; these ranges can be maintained up to 15 hours.
  • Maintain specific temperature ranges for extended periods of time for improved sample protection
  • Get the temperature protection you need with choice of durable transport bags or boxes
  • Modular, color-coded cooling packs allow flexible configurations for different-sized samples
  • Ensure sample integrity with optional security sealing ties
Specifications
Specifications
1/Pk.
2°C to 8°C for 8 to 15 hours
For transporting temperature sensitive samples
Blue Cooling Block (2 L)