Learn More
Cole-Parmer™ PolarSafe™ Cooling Packs and Accessories
Transport your critical samples without worry
Specifications
Applications For transporting temperature sensitive samples
View More Specs
Products 11
Catalog Number Description Temperature Range (Metric) Quantity Price Quantity & Availability  
Catalog Number Description Temperature Range (Metric) Quantity Price Quantity & Availability  
13-200-150
View Documents
Blue Cooling Block (1 L) 2°C to 8°C for 8 to 15 hours 1/Pk. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-200-151
View Documents
Blue Slim Cooling Block (1 L) 2°C to 8°C for 8 to 15 hours 1/Pk. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-200-152
View Documents
Blue Cooling Block (2 L) 2°C to 8°C for 8 to 15 hours 1/Pk. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-200-148
View Documents
Green Cooling Block (1 L) 20°C to 24°C for 8 to 15 hours 1/Pk. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-200-149
View Documents
Green Slim Cooling Block (1 L) 20°C to 24°C for 8 to 15 hours 1/Pk. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-200-153
View Documents
Green Cooling Block (2 L) 20°C to 24°C for 8 to 15 hours 1/Pk. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-200-146
View Documents
Blue Cooling End-caps 2°C to 8°C for 8 to 15 hours 2/Pk. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-200-144
View Documents
Green Cooling End-caps 20°C to 24°C for 8 to 15 hours 2/Pk. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-200-147
View Documents
Blue Cooling Frames 2°C to 8°C for 8 to 15 hours 2/Pk. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-200-145
View Documents
Green Cooling Frames 20°C to 24°C for 8 to 15 hours 2/Pk. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-200-154
View Documents
Security Sealing Tie 100/Cs. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Use the cooling end-caps and cooling frames together to encapsulate the entire sample.
  • This modular system can accommodate various sample sizes to provide a complete thermal barrier.
  • Temperature ranges are 2 to 8°C and 20 to 24°C respectively; these ranges can be maintained up to 15 hours.
  • Maintain specific temperature ranges for extended periods of time for improved sample protection
  • Get the temperature protection you need with choice of durable transport bags or boxes
  • Modular, color-coded cooling packs allow flexible configurations for different-sized samples
  • Ensure sample integrity with optional security sealing ties
Specifications
Specifications
For transporting temperature sensitive samples