Learn More
Cole-Parmer™ PolarSafe™ Cooling Transport Bags
Transport your critical samples without worry
 View more versions of this product
Catalog No. 13-200-136
Add to cart
Description
Description
  • Insulated nylon bags provide an easy solution for transporting your samples.
  • Bags have pockets to keep documentation safe during transit, a shoulder strap, and carrying handles.
  • Maintain specific temperature ranges for extended periods of time for improved sample protection
  • Get the temperature protection you need with choice of durable transport bags or boxes
  • Modular, color-coded cooling packs allow flexible configurations for different-sized samples
  • Ensure sample integrity with optional security sealing ties
Specifications
Specifications
For transporting temperature sensitive samples
30 L
Transport Bag with Two 4°C Blocks (1 L, Slim) and Four 4°C Blocks (1 L)