Learn More
Cole-Parmer™ PolarSafe™ Cooling Transport Boxes
Transport your critical samples without worry
 View more versions of this product
Catalog No. 13-200-139
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Durable polypropylene-insulated walls offer improved insulation for better sample protection.
  • Boxes are stackable and feature a convenient shoulder strap and integrated handle.
  • Maintain specific temperature ranges for extended periods of time for improved sample protection
  • Get the temperature protection you need with choice of durable transport bags or boxes
  • Modular, color-coded cooling packs allow flexible configurations for different-sized samples
  • Ensure sample integrity with optional security sealing ties
Specifications
Specifications
5 L
290 x 170 x 130 mm
Transport Box with Two 22°C End-Caps and Two 22°C Frames
For transporting temperature sensitive samples