Learn More
Cole-Parmer™ PolarSafe™ Cooling Transport Boxes
Transport your critical samples without worry
$194.00 - $673.00
Specifications
Applications For transporting temperature sensitive samples
View More Specs
Products 9
Catalog Number Capacity (Metric) Includes Price Quantity & Availability  
Catalog Number Capacity (Metric) Includes Price Quantity & Availability  
13-200-127
View Documents
Transport Box Each for $194.00
Add to cart
13-200-138
View Documents
Transport Box with Two 4°C End-Caps and Two 4°C Frames Each for $488.00
Add to cart
13-200-139
View Documents
Transport Box with Two 22°C End-Caps and Two 22°C Frames Each for $488.00
Add to cart
13-200-128
View Documents
Transport Box Each for $230.00
Add to cart
13-200-140
View Documents
Transport Box with Two 4°C End-Caps and Four 4°C Frames Each for $673.00
Add to cart
13-200-141
View Documents
Transport Box with Two 22°C End-Caps and Four 22°C Frames Each for $673.00
Add to cart
13-200-129
View Documents
Transport Box Each for $267.00
Add to cart
13-200-142
View Documents
Transport Box with Two 4°C Blocks (2 L) Each for $534.00
Add to cart
13-200-143
View Documents
Transport Box with Two 22°C Blocks (2 L) Each for $534.00
Add to cart
Description
Description
  • Durable polypropylene-insulated walls offer improved insulation for better sample protection.
  • Boxes are stackable and feature a convenient shoulder strap and integrated handle.
  • Maintain specific temperature ranges for extended periods of time for improved sample protection
  • Get the temperature protection you need with choice of durable transport bags or boxes
  • Modular, color-coded cooling packs allow flexible configurations for different-sized samples
  • Ensure sample integrity with optional security sealing ties
Specifications
Specifications
For transporting temperature sensitive samples