Learn More
Argos Technologies™ PolarSafe™ Cryo/Freezer Cell Dividers Only For Standard Tubes
Designed only for standard tubes. Argos Technologies PolarSafe™ Cryo/Freezer Cell Dividers Only For Standard Tubes has small holes on bottom which allows safe drainage of liquid nitrogen or water.
$38.90 - $47.15
Specifications
Type Cell Divider
For Use With (Application) Sample Storage for Laboratories in Universities and for Performing Artificial Insemination, Blood Bank, and Stem Cell/Bone Marrow Procedures in Institutions
Material Fiberboard
Quantity 12 Pack
Color White
View More Specs
Products 7
Catalog Number Description Diameter (Metric) Well Dimensions (L x W x H) Price Quantity & Availability  
Catalog Number Description Diameter (Metric) Well Dimensions (L x W x H) Price Quantity & Availability  
03-391-511
View Documents
81-place cell divider 12.7 mm 12.1 x 12.1 x 3.3 cm (4.8 x 4.8 x 1.3 in.)
Pack of 12 for $39.70
Add to cart
03-391-509
View Documents
49-place cell divider 16.7 mm 12.1 x 12.1 x 3.3 cm (4.8 x 4.8 x 1.3 in.)
Pack of 12 for $39.70
Add to cart
03-391-506
View Documents
25-place cell divider 11.5 mm 2.6 x 2.6 x 3 cm (2.6 x 2.6 x 1.2 in.)
Pack of 12 for $47.15
Add to cart
03-391-507
View Documents
25-place cell divider 24.5 mm 12.1 x 12.1 x 3.3 cm (4.8 x 4.8 x 1.3 in.)
Pack of 12 for $39.70
Add to cart
03-391-510
View Documents
64-place cell divider 14.6 mm 12.1 x 12.1 x 3.3 cm (4.8 x 4.8 x 1.3 in.)
Pack of 12 for $39.70
Add to cart
03-391-512
View Documents
100-place cell divider 11.5 mm 12.1 x 12.1 x 3.3 cm (4.8 x 4.8 x 1.3 in.)
Pack of 12 for $39.70
Add to cart
03-391-513
View Documents
144-place cell divider 9.5 mm 12.1 x 12.1 x 3.3 cm (4.8 x 4.8 x 1.3 in.)
Pack of 12 for $38.90
Add to cart
Description
Description
  • Made of white fiberboard to protect samples
  • Accommodates range of tube sizes from 8mm to 33.5mm
  • Dividers can hold 16 to 196 tubes
  • Special moisture-repellent coating protects effects of liquid nitrogen or codensation from refrigerated equipment
Specifications
Specifications
Cell Divider
Fiberboard
White
Sample Storage for Laboratories in Universities and for Performing Artificial Insemination, Blood Bank, and Stem Cell/Bone Marrow Procedures in Institutions
12 Pack