Learn More
Argos Technologies™ PolarSafe™ Cryo/Freezer Standard Cardboard Box With Dividers
Used in mechanical freezers and liquid nitrogen tanks. Argos Technologies PolarSafe™ Cryo/Freezer Standard Cardboard Box With Dividers has small holes on bottom which allows safe drainage of liquid nitrogen or water.
$7.90 - $7.95
Specifications
Type Standard Cardboard Box With Dividers
For Use With (Application) Sample storage for laboratories in universities and for performing artificial insemination, blood bank, and stem cell/bone marrow procedures in institutions
Material Fiberboard
Dimensions (L x W x H) 13.2 x 13.2 x 5 cm (5.2 x 5.2 x 2 in.)
Color White
View More Specs
Products 2
Catalog Number Description Price Quantity & Availability  
Catalog Number Description Price Quantity & Availability  
03-391-515 View Documents 81-place cell divider Each for $7.90
Add to cart
03-391-516 View Documents 100-place cell divider Each for $7.95
Add to cart
Description
Description
  • Made of white fiberboard to protect samples
  • Special moisture-repellent coating protects effects of liquid nitrogen or condensation from refrigerated equipment
  • Surface is easy to write on for identification
Specifications
Specifications
Standard Cardboard Box With Dividers
Fiberboard
White
Sample storage for laboratories in universities and for performing artificial insemination, blood bank, and stem cell/bone marrow procedures in institutions
13.2 x 13.2 x 5 cm (5.2 x 5.2 x 2 in.)