Learn More
Argos Technologies PolarSafe™ Dots for Cryo Storage
Use in temperatures as low as -196°C (-320°F)
Manufacturer:  Argos Technologies L1291
 View more versions of this product
Catalog No. 03-391-111
  Check Availability
Add to cart
Description
Description
  • Suitable for use on microcentrifuge tubes, cryovials, racks, boxes, and microtiter plates
  • No need to self-laminate
  • Thinner labels mean tubes will still fit most storage systems
  • Easy to see—bold colors let you identify samples while in the box
  • Unique adhesive formulation for extra staying power
Order Info
Available on rolls or sheets for use with laser printers.
Specifications
Specifications
White
Self-adhesive labels designed specifically for low temperature use
-320.8°F
0.75 in.
Polyester
192 Sheet(s)
19mm
2.0 to 5.0mL tubes
-196°C
Label
Micryo Dot