Argos Technologies PolarSafe™ ICE Buckets and Pans

Ice buckets and pans have excellent insulating properties to help keep temperature sensitive samples safely chilled.

Manufacturer: Argos Technologies B8801BL

 View more versions of this product

Catalog No. 10-987-062

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Capacity (English) 0.26 gal
    For Use With (Application) Ice, dry ice, slurry ice
    Material Polystyrene
    Color Black
    Capacity (Metric) 1L
    Product Type Ice pan

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit