Learn More
Argos Technologies™ PolarSafe™ ICE Buckets and Pans
Ice buckets and pans have excellent insulating properties to help keep temperature sensitive samples safely chilled.
Description
- High density expanded polystyrene construction provides extra rigidity and strength, and to prevent condensation on benchtops
- Buckets come with recessed handle and snug fitting lids with easy to grab knob to minimize evaporation loss and to maximize cooling
- Pans have gripping ridges for easy transportation and pour spout to empty cooling material when no longer needed
Specifications
0.66 gal. | |
Black | |
Lid | |
Ice Bucket |
2.5 L | |
Ice, dry ice, slurry ice | |
Polystyrene |