Learn More
Argos Technologies™ PolarSafe™ ICE Buckets and Pans
Ice buckets and pans have excellent insulating properties to help keep temperature sensitive samples safely chilled.
 View more versions of this product
Catalog No. 10-987-064
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • High density expanded polystyrene construction provides extra rigidity and strength, and to prevent condensation on benchtops
  • Buckets come with recessed handle and snug fitting lids with easy to grab knob to minimize evaporation loss and to maximize cooling
  • Pans have gripping ridges for easy transportation and pour spout to empty cooling material when no longer needed
Specifications
Specifications
Green
Ice Pan
1.98 gal.
Polystyrene
7.5 L
Ice, dry ice, slurry ice