Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Argos Technologies PolarSafe™ Strips for Cryo Storage

Use in temperatures as low as -196°C (-320°F)

Manufacturer:  Argos Technologies L1281

 View more versions of this product

Catalog No. 03-391-108


Add to cart

Description

Description

Use in temperatures as low as -196°C (-320°F)

  • Suitable for use on microcentrifuge tubes, cryovials, racks, boxes, and microtiter plates
  • No need to self-laminate
  • Thinner labels mean tubes will still fit most storage systems
  • Easy to see—bold colors let you identify samples while in the box
  • Unique adhesive formulation for extra staying power

Order Info

Available on rolls or sheets for use with laser printers.

Specifications

Specifications

White
0.94 x 0.5 in.
-320.8°F
Label
Micryo Strip
0.5mL Tube, laser printer, can even be used in incubators
2.4 x 1.3cm
-196°C
Polyester
119 Sheet(s)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit