Learn More
Argos Technologies PolarSafe™ Strips for Cryo Storage
Use in temperatures as low as -196°C (-320°F)
Manufacturer:  Argos Technologies L1285
 View more versions of this product
Catalog No. 03-391-113
  Check Availability
Add to cart
Description
Description
Use in temperatures as low as -196°C (-320°F)
  • Suitable for use on microcentrifuge tubes, cryovials, racks, boxes, and microtiter plates
  • No need to self-laminate
  • Thinner labels mean tubes will still fit most storage systems
  • Easy to see—bold colors let you identify samples while in the box
  • Unique adhesive formulation for extra staying power
Order Info
Available on rolls or sheets for use with laser printers.
Specifications
Specifications
Rainbow
1.5 x 0.25 in.
-320.8°F
Label
Micryo Strip
96 and 384-well plates, laser printer, can even be used in incubators
3.8 x 0.6cm
-196°C
Polyester
156 Sheet(s)