Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLD3 Rabbit anti-Human, Polyclonal , Abnova™

POLD3 polyclonal antibody raised against recombinant human POLD3.

Manufacturer:  Abnova Corporation PAB29475

Catalog No. 89-934-790


Add to cart

Description

Description

The DNA polymerase delta complex is involved in DNA replication and repair, and it consists of the proliferating cell nuclear antigen (PCNA; MIM 176740), the multisubunit replication factor C (see MIM 102579), and the 4 subunit polymerase complex: POLD1 (MIM 174761), POLD2 (MIM 600815), POLD3, and POLD4 (MIM 611525) (Liu and Warbrick, 2006 [PubMed 16934752]).[supplied by OMIM]

Sequence: SVTEPKLATPAGLKKSSKKAEPVKVLQKEKKRGKRVALSDDETKETENMRKKRRRIKLPESDSSEDEVFPDSPGAY
Specifications

Specifications

POLD3
Unconjugated
Immunofluorescence (1-4 ug/mL) Immunohistochemistry (1:500-1:1000) The optimal working dilution should be determined by the end user.
Liquid
POLD3
Recombinant protein corresponding to amino acids of human POLD3.
Antigen affinity purification
RUO
Primary
10714
Immunofluorescence, Immunohistochemistry (PFA fixed)
POLD3 polyclonal antibody raised against recombinant human POLD3.
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
KIAA0039/MGC119642/MGC119643/P66/P68
Rabbit
IgG
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit