Learn More
POLDIP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 150801AP
Catalog No. 50-172-9960
Add to cart
Description
Description
POLDIP2 is a protein that interacts with the DNA polymerase delta p50 subunit, as well as with proliferating cell nuclear antigen. The encoded protein maybe play a role in the ability of the replication fork to bypass DNA lesions. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
POLDIP2
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q91VA6, Q9Y2S7
PDIP38, POLDIP2
Rabbit
IgG
150 μL
-20°C
Primary
26073, 287544, 67811
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
POLDIP2
Liquid
Poldip2
POLDIP2 Fusion Protein Ag7154
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat