Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poliovirus 3 Mouse anti-Virus, Alexa Fluor™ 350, Clone: 4D5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223575AF350

Catalog No. NP2237A30


Add to cart

Description

Description

Poliovirus 3 Monoclonal antibody specifically detects Poliovirus 3 in Virus samples. It is validated for ELISA.
Specifications

Specifications

Poliovirus 3
4D5
ELISA
Conjugated, Purified
Poliovirus type 3 (inactivated)
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Virus
ELISA
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG2a Kappa
0.1 mL
Primary
There is no cross reactivity with type 1 and 2.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit