Learn More
Poliovirus 3 Mouse anti-Virus, Alexa Fluor™ 405, Clone: 4D5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP223575AF405
Catalog No. NP2237A40
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Poliovirus 3 Monoclonal antibody specifically detects Poliovirus 3 in Virus samples. It is validated for ELISA.
Specifications
Specifications
Poliovirus 3
4D5
ELISA
Mouse
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
Virus
ELISA
Alexa Fluor™ 405
50 mM sodium borate with 0.05% sodium azide
Poliovirus type 3 (inactivated)
0.1 mL
Primary
There is no cross reactivity with type 1 and 2.