Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLN Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548705
Description
This gene encodes a DNA polymerase type-A family member. The encoded protein plays a role in DNA repair and homologous recombination. This gene shares its 5′ exons with some transcripts from overlapping GeneID: 79441, which encodes an augmentin-like protein complex subunit. [provided by RefSeq, Dec 2014]Specifications
POLN | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q7TQ07 | |
POLN | |
KLH conjugated synthetic peptide between 651-679 amino acids from the C-terminal region of human POLN. | |
400 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
Poln | |
DNA polymerase nu, POLN | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
272158 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction