Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Poloxamer 188

Click to view available options
Quantity:
500g
1kg
9003-11-6
C5H10O2
102.13
MFCD00082049
RVGRUAULSDPKGF-UHFFFAOYNA-N
Pluronic™ F-68; Polyoxyethylene-polyoxypropylene block copolymer
24751
2-methyloxirane;oxirane
CC1CO1.C1CO1
9003-11-6
C5H10O2
102.13
MFCD00082049
RVGRUAULSDPKGF-UHFFFAOYNA-N
Pluronic™ F-68; Polyoxyethylene-polyoxypropylene block copolymer
24751
2-methyloxirane;oxirane
CC1CO1.C1CO1
Applications
Poloxamer is a non-ionic detergent that protects cells from hydrodynamic damage. It can be also used for in vitro isolation of lymphocytes from diluted whole blood.

Solubility
Miscible with water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Linear Formula (C3H6O·C2H4O)x
Physical Form Solid
Color White
Quantity 1kg
Chemical Name or Material Poloxamer 188

RTECSNumber : MD0911050

TSCA : TSCA