Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLR1B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548383

Catalog No. PA548383


Add to cart

Description

Description

Predicted to react with rat based on sequence homology.

DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Second largest core component of RNA polymerase I which synthesizes ribosomal RNA precursors. Proposed to contribute to the polymerase catalytic activity and forms the polymerase active center together with the largest subunit. Pol I is composed of mobile elements and RPA2 is part of the core element with the central large cleft and probably a clamp element that moves to open and close the cleft.
Specifications

Specifications

POLR1B
Polyclonal
PBS with 0.09% sodium azide
Q9H9Y6
DNA-directed RNA polymerase I subunit RPA2, RNA polymerase I subunit 2, DNA-directed RNA polymerase I 135 kDa polypeptide, RPA135, POLR1B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84172
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
POLR1B
Liquid
POLR1B
KLH conjugated synthetic peptide between 89-117 amino acids from the N-terminal region of human POLR1B.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit