Learn More
Aviva Systems Biology Corp POLR1D Antibody - middle region (ARP48787_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48787_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POLR1D. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human POLR1D. Alternate names: FLJ20616, MGC9850, POLR1C, RPA16, RPA9, RPAC2, RPO1-3, AC19, TCS2, RPC16ApplicationWB. Species reactivity: Cow, Dog, Human, Rabbit
Catalog No. 50-014-59922