Learn More
POLR1E Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 161451AP
Catalog No. 50-173-0751
Add to cart
Description
Description
The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae. Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. This gene encodes a member of the golgin family of proteins which are localized to the Golgi. Its encoded protein has been postulated to play a role in nuclear transport and Golgi apparatus localization. Several alternatively spliced transcript variants that encode different protein isoforms have been described for this gene.
Specifications
Specifications
POLR1E
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8K202, Q9GZS1
PAF53, POLR1E, PRAF1, RNA polymerase I subunit A49, RP11 405L18.3
Rabbit
IgG
150 μL
-20°C
Primary
313245, 64424, 64425
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
POLR1E
Liquid
Polr1e
POLR1E Fusion Protein Ag9269
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat