Learn More
POLR2F Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 153341AP
Catalog No. 50-173-0156
Add to cart
Description
Description
POLR2F belongs to the archaeal rpoK/eukaryotic RPB6 RNA polymerase subunit family and is the sixth largest subunit of RNA polymerase II. This polymerase, along with two other DNA-directed RNA polymerases, is responsible for synthesizing messenger RNA in eukaryotes. Three distinct zinc-containing RNA polymerases are found in eukaryotic nuclei: polymerase I for the ribosomal RNA precursor, polymerase II for the mRNA precursor, and polymerase III for 5S and tRNA genes. Each class of RNA polymerase is assembled from 9 to 15 different polypeptides. In yeast, this polymerase subunit, in combination with at least two other subunits, forms a structure that stabilizes the transcribing polymerase on the DNA template.
Specifications
Specifications
POLR2F
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O88828, P61218, P61219
HRBP14.4, POLR2F, POLRF, RPABC14.4, RPABC2, RPB14.4, RPB6, RPB6 homolog, RPC15
Rabbit
IgG
150 μL
-20°C
Primary
5435, 69833, 83503
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
POLR2F
Liquid
POLR2F
POLR2F Fusion Protein Ag7449
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat