Learn More
Aviva Systems Biology Corp POLR2H Antibody - N-terminal region (ARP48697_T100) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48697_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against POLR2H. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human POLR2H. Alternate names: RPABC3, RPB17, RPB8, hsRPB8ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59888