Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLR2J2 Mouse anti-Human, HRP, Clone: OTI3G4, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273520H
Description
POLR2J2 Monoclonal specifically detects POLR2J2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
POLR2J2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
POLR2J2 | |
Full length recombinant protein of human POLR2J2 (NP_116581) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI3G4 | |
Western Blot, Immunohistochemistry | |
DNA directed RNA polymerase II polypeptide J-related, DNA-directed RNA polymerase II subunit RPB11-b1, HRPB11B, MGC105050, MGC54043, polymerase (RNA) II (DNA directed) polypeptide J2, RNA polymerase II subunit B11-b1, RPB11, RPB11b1DNA-directed RNA polymerase II subunit J2 | |
Mouse | |
Affinity Purified | |
DNA replication Transcription Translation and Splicing | |
246721.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title