Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLR3A Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | POLR3A |
---|---|
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB022979
|
Novus Biologicals
NBP294211100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB022980
|
Novus Biologicals
NBP29421120UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
POLR3A Polyclonal antibody specifically detects POLR3A in Human, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
POLR3A | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.3), 50% glycerol | |
11128 | |
IgG | |
Affinity purified |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Rat | |
DNA-directed RNA polymerase III largest subunit, DNA-directed RNA polymerase III subunit A, DNA-directed RNA polymerase III subunit RPC1, EC 2.7.7.6, hRPC155, POL3A, polymerase (RNA) III (DNA directed) polypeptide A, 155kDa, RNA polymerase III 155 kDa subunit, RNA polymerase III subunit C160, RNA polymerase III subunit RPC155-D, RPC1, RPC155RNA polymerase III subunit C1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human POLR3A (NP_008986.2). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title