Learn More
Aviva Systems Biology Corp POLR3F Antibody - N-terminal region (ARP48704_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48704_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POLR3F. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human POLR3F. Alternate names: MGC13517, RPC39, RPC6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59891