Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POLR3GL Monoclonal Antibody (OTI5F9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526289
Description
POLR3GL Monoclonal Antibody for Western Blot, ICC/IF
DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Specific peripheric component of RNA polymerase III which synthesizes small RNAs, such as 5S rRNA and tRNAs.
Specifications
POLR3GL | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BT43 | |
POLR3GL | |
Full length human recombinant protein of POLR3GL produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot, Immunocytochemistry | |
OTI5F9 | |
Unconjugated | |
POLR3GL | |
2010003I19Rik; alternative RNA polymerase III subunit 32; DNA-directed RNA polymerase III subunit G-like; DNA-directed RNA polymerase III subunit RPC7-like; flj32422; POLR3GL; polymerase (RNA) III (DNA directed) polypeptide G (32kD)-like; polymerase (RNA) III (DNA directed) polypeptide G like; polymerase (RNA) III subunit G like; RNA polymerase III 32 kDa beta subunit; RNA polymerase III subunit C7-like; RNA polymerase III subunit G like; RPC32-beta; RPC32HOM; RPC32-like protein | |
Mouse | |
Affinity chromatography | |
RUO | |
84265 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction