Learn More
POLR3K 293T Cell Overexpression Lysate 2 (Denatured), Abnova
Manufacturer:  Abnova Corporation H00051728T02
Catalog No. 89-002-257
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
Western Blot
293T
NM_016310.2
6.8
Transient overexpression cell Lysate was tested with Anti-POLR3K antibody (H00051728-B02) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
POLR3K 293T Cell Transient Overexpression Lysate(Denatured)
11.99
pCMV-POLR3K full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.