Learn More
Aviva Systems Biology Corp TENT4A Antibody - N-terminal region (ARP48720_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48720_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POLS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human POLS. Alternate names: LAK-1, POLK, TRF4, TRF4-1, LAK1, POLS, TRF41, TUTASE5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59899