Poltex Gel Stain Boxes and Lids
Can be used for light sensitive stains (like SYBR Green or equivalent), or for light sensitive incubations
Manufacturer: POLTEX LLC BT01019BL
 View more versions of this product
Catalog No. 22-000-178
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    For Use With (Equipment) Gel Stain Box
    Material Polycarbonate
    Product Type Lid for large black polycarbonate gel stain box (BT-01019B)
  • Description & Specifications
Related Products
Product Content Feedback