Learn More
Poltex In-Hood Serological Pipet Holder
Holder allows storage of pipettes inside tissue culture hood
Manufacturer:  POLTEX LLC LO01002A
 View more versions of this product
Catalog No. 22-000-190
  Check Availability
Add to cart
Specifications
Description
  • Storage inside tissue culture hood maximize sterile technique and ergonomics
  • Additional items can be stored on the flat top
  • 10 in. depth allows for better maneuverability inside hood
  • Shelves are removable for easy cleaning
  • Available with or without side pockets
  • Pocket dimensions (L x W x H): 25.4 x 6.35 x 12.7cm (10 x 2.5 x 5 in.)
Description & Specifications
Specifications
Holder, Pipette
Base Model
25.4 x 14.6 x 26.67cm (10 x 5.75 x 10.5 in.)