Learn More
Poltex ST Glove Box Holders
Position vertically or horizontally to change out glove boxes with ease.
$55.00 - $108.10
Specifications
Material Polycarbonate
Product Type Glove box holder
View More Specs
Products 8
Catalog Number Mfr. No. No. of compartments Mounting Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. No. of compartments Mounting Width (English) Price Quantity & Availability  
22-000-130
View Documents
Poltex
1GBSTW
1 Pre-drilled holes 6 in. Each for $55.00
Add to cart
22-000-131
View Documents
Poltex
1GBSTM
1 Magnets 6 in. Each for $68.80
Add to cart
22-000-134
View Documents
Poltex
2GBSTW
2 Pre-drilled holes 11.5 in. Each for $69.80
Add to cart
22-000-135
View Documents
Poltex
2GBSTM
2 Magnets 11.5 in. Each for $84.10
Add to cart
22-000-139
View Documents
Poltex
3GBSTM
3 Magnets 17.25 in. Each for $92.00
Add to cart
22-000-138
View Documents
Poltex
3GBSTW
3 Pre-drilled holes 17.25 in. Each for $78.10
Add to cart
22-000-142
View Documents
Poltex
4GBSTW
4 Pre-drilled holes 22.75 in. Each for $94.10
Add to cart
22-000-143
View Documents
Poltex
4GBSTM
4 Magnets 22.75 in. Each for $108.10
Add to cart
Description
Description
  • Choose from 1 to 4 boxes of gloves
  • Strong polycarbonate construction that is highly break resistant
  • 10 in. tall
Mounting options for holders:
  • Pre-drilled holes for wall mounting
  • Powerful magnets that securely mount the holder to ay metal surface
Specifications
Specifications
Polycarbonate
Glove box holder