Learn More
Poltex Blood Tube Rack Organizer Tray
Provides safe transport of blood tubes throughout clinical lab
Manufacturer:  POLTEX LLC LO130504
 View more versions of this product
Catalog No. 22-000-170
  Check Availability
Add to cart
Specifications
Description
  • Constructed of super durable, break-resistant PETG
  • Easily and economically customizable
  • Choice of 3 or 4 rack capacity
  • Pocket size (D x W x H): 10.4 x 24.7 x 5.08cm (4.125 x 9.75 x 2 in.)
  • 2 in. tall sidewalls
  • Cleanable with 70% Isopropyl Alcohol
Description & Specifications
Specifications
Polycarbonate
Tray
22.35 x 50.8cm (8.8 x 20 in.)
Blood Sample Tubes