Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(2-acrylamido-2-methyl-1-propanesulfonic acid), 10 wt% aq.sol.; ca. MW 800,000, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
27119-07-9,7732-18-5
C7H13NO4S
207.244
MFCD00084369
XHZPRMZZQOIPDS-UHFFFAOYSA-N
2-acrylamido-2-methylpropanesulfonic acid,2-acrylamido-2-methyl-1-propanesulfonic acid,2-acrylamide-2-methylpropanesulfonic acid,1-propanesulfonic acid, 2-methyl-2-1-oxo-2-propenyl amino,polyacrylamidomethylpropane sulfonic acid,unii-490hqe5ki5,2-acrylamido-2-methylpropanesulfonate,2-acrylamido-2-methylpropanesulphonic acid,2-acrylamido-2-methylpropane sulfonic acid,2-acrylamido-2-methylpropane-1-sulfonic acid
65360
2-methyl-2-(prop-2-enoylamino)propane-1-sulfonic acid
CC(C)(CS(=O)(=O)O)NC(=O)C=C
27119-07-9,7732-18-5
C7H13NO4S
207.244
MFCD00084369
XHZPRMZZQOIPDS-UHFFFAOYSA-N
2-acrylamido-2-methylpropanesulfonic acid,2-acrylamido-2-methyl-1-propanesulfonic acid,2-acrylamide-2-methylpropanesulfonic acid,1-propanesulfonic acid, 2-methyl-2-1-oxo-2-propenyl amino,polyacrylamidomethylpropane sulfonic acid,unii-490hqe5ki5,2-acrylamido-2-methylpropanesulfonate,2-acrylamido-2-methylpropanesulphonic acid,2-acrylamido-2-methylpropane sulfonic acid,2-acrylamido-2-methylpropane-1-sulfonic acid
65360
2-methyl-2-(prop-2-enoylamino)propane-1-sulfonic acid
CC(C)(CS(=O)(=O)O)NC(=O)C=C
Linear Formula {-CH2CH[CONHC(CH3)2CH2SO3H]-}n
Physical Form Viscous Liquid
Infrared Spectrum Authentic
Percent Purity 8.0 to 12.0% (solids)
Packaging Glass bottle
Color Undesignated
Quantity 25g
Chemical Name or Material Poly(2-acrylamido-2-methyl-1-propanesulfonic acid), 10 wt% aqueous solution

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning