Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(2-acrylamido-2-methyl-1-propanesulfonic acid-co-styrene), sodium salt, styrene content 95%, M.W. ∽ 500,000, Acros Organics

Click to view available options
Quantity:
10g
Packaging:
Plastic bottle
51121-85-8
C15H21NO4S
311.396
MFCD00084371
UHBZMSVSQFNVRN-UHFFFAOYSA-N
1-propanesulfonic acid, 2-methyl-2-1-oxo-2-propenyl amino-, polymer with ethenylbenzene,styrene 2-acrylamido-2-methyl-1-propanesulfonic acid,2-methyl-2-prop-2-enamido propane-1-sulfonic acid; styrene,2-methyl-2-prop-2-enoylamino propane-1-sulfonic acid; styrene
3084964
2-methyl-2-(prop-2-enoylamino)propane-1-sulfonic acid;styrene
CC(C)(CS(=O)(=O)O)NC(=O)C=C.C=CC1=CC=CC=C1
51121-85-8
C15H21NO4S
311.396
MFCD00084371
UHBZMSVSQFNVRN-UHFFFAOYSA-N
1-propanesulfonic acid, 2-methyl-2-1-oxo-2-propenyl amino-, polymer with ethenylbenzene,styrene 2-acrylamido-2-methyl-1-propanesulfonic acid,2-methyl-2-prop-2-enamido propane-1-sulfonic acid; styrene,2-methyl-2-prop-2-enoylamino propane-1-sulfonic acid; styrene
3084964
2-methyl-2-(prop-2-enoylamino)propane-1-sulfonic acid;styrene
CC(C)(CS(=O)(=O)O)NC(=O)C=C.C=CC1=CC=CC=C1
Physical Form Crystalline Powder
Infrared Spectrum Authentic
Loss on Drying 5% max.
Packaging Plastic bottle
Color White
Quantity 10g
Chemical Name or Material Poly(2-acrylamido-2-methyl-1-propanesulfonic acid-co-styrene), sodium salt

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.