Alfa Aesar™ Poly(2-methylaniline), emeraldine salt from p-toluenesulfonic acid

Manufacturer: Alfa Aesar™ 4380614

Quantity 25g

 View more versions of this product

Catalog No. AA4380614

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula −[C6H3(CH3)NH-]n·(C6H4CH3SO3)m
    MDL Number MFCD02688597
    Chemical Name or Material Poly (2-methylaniline), Emeraldine salt from p-toluenesulfonic acid

    Specifications

    Physical Form Powder
    Molecular Formula −[C6H3(CH3)NH-]n·(C6H4CH3SO3)m
    Quantity 25g
    MDL Number MFCD02688597
    Chemical Name or Material Poly (2-methylaniline), Emeraldine salt from p-toluenesulfonic acid

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit