Alfa Aesar™ Poly(3-butylthiophene-2,5-diyl), regiorandom

Manufacturer: Alfa Aesar™ 4478803

Quantity 1g

 View more versions of this product

Catalog No. AA4478803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 98837-51-5
    Molecular Formula (C8H12S)n
    MDL Number MFCD00677712
    Chemical Name or Material Poly (3-butylthiophene-2,5-diyl), Regiorandom

    Specifications

    Physical Form Powder
    CAS 98837-51-5
    Quantity 1g
    Molecular Formula (C8H12S)n
    MDL Number MFCD00677712
    Color Brown to black
    Chemical Name or Material Poly (3-butylthiophene-2,5-diyl), Regiorandom

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit