Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(4-bromostyrene), average M.W. 65000 (GPC), Acros Organics

Click to view available options
Quantity:
1g
24936-50-3
24936-50-3
Linear Formula {-CH2CH(C6H4Br)-}n
Physical Form Powder or Chunks
Infrared Spectrum Authentic
Color White
Quantity 1g
Chemical Name or Material Poly(4-bromostyrene)