Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(4-methoxystyrene), ∽ M.W. 400,000, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
24936-44-5
MFCD00134007
24936-44-5
MFCD00134007
Linear Formula {-CH2CH(C6H4OCH3)-}n
Physical Form Powder
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Quantity 1g
Chemical Name or Material Poly(4-methoxystyrene)