Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(4-methyl-1-pentene), low molecular weight, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
25068-26-2
C6H12
84.162
MFCD00084382
WSSSPWUEQFSQQG-UHFFFAOYSA-N
4-methyl-1-pentene,1-pentene, 4-methyl,poly 4-methyl-1-pentene,unii-x10hrj2y7w,x10hrj2y7w,poly 4-methylpentene,isobutylethene,isohexene,4-methylpentene-1,1-pentene, 4-methyl-, homopolymer
12724
4-methylpent-1-ene
CC(C)CC=C
25068-26-2
C6H12
84.162
MFCD00084382
WSSSPWUEQFSQQG-UHFFFAOYSA-N
4-methyl-1-pentene,1-pentene, 4-methyl,poly 4-methyl-1-pentene,unii-x10hrj2y7w,x10hrj2y7w,poly 4-methylpentene,isobutylethene,isohexene,4-methylpentene-1,1-pentene, 4-methyl-, homopolymer
12724
4-methylpent-1-ene
CC(C)CC=C
Density 0.83g/mL
Linear Formula {-CH2CHCH2CH(CH3)2-}n
Physical Form Beads
Infrared Spectrum Authentic
Loss on Drying 1% max.
Packaging Glass bottle
Color Undesignated
Quantity 500g
Chemical Name or Material Poly(4-methyl-1-pentene)