Alfa Aesar™ Poly(4-vinylpyridine), 2% cross-linked, ca 60 mesh, 8.0meq/g

Manufacturer: Alfa Aesar™ B202790B

Quantity 1000g

 View more versions of this product

Catalog No. AAB202790B

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 9017-40-7
    MDL Number MFCD00190588
    Chemical Name or Material Poly(4-vinylpyridine), 2% cross-linked

    Specifications

    TSCA TSCA
    MDL Number MFCD00190588
    Odor Odorless
    Concentration 8.0mEq/g
    Quantity 1000g
    Mesh Size ≈60 mesh
    CAS 9017-40-7
    Chemical Name or Material Poly(4-vinylpyridine), 2% cross-linked

    Description

    Applications
    Useful as an acid scavenger in non-aqueous systems, e.g., in the reaction of acid halides with alcohols or amines, as catalyst in Knoevenagel-type condensations and for the formation of enamines and ketals, replacement for pyridine, enabling simplified workup and product isolation.

    Solubility
    Insoluble in water.

    Notes
    Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit