Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(4-vinylpyridine), 2% cross-linked with DVB, ca. 60 mesh, ACROS Organics™

Click to view available options
Quantity:
50g
CAS 9017-40-7
Additional Information base capacity: ca. 8.0 meq/g (dry)
Molecular Formula C17H17N
MDL Number MFCD00190588
Fieser 16,282
Synonym 4-vinylpyridine; divinylbenzene,diethenylbenzene, polymer with 4-ethenylpyridine,pyridine, 4-ethenyl-, polymer with diethenylbenzene,4-vinylpyridine divinylbenzene polymer,4-vinylpyridine divinylbenzene,1,2-divinylbenzene; 4-vinylpyridine,poly 4-vinylpyridine cross-linked,1,2-bis ethenyl benzene; 4-ethenylpyridine,1,2-diethenylbenzene-4-ethenylpyridine 1:1,poly 4-vinylpyridine cross-linked, ca 60 mesh, 8.0meq/g
InChI Key HUEZUDXJCGHIHG-UHFFFAOYSA-N
SMILES C=CC1=CC=NC=C1.C=CC1=CC=CC=C1C=C
IUPAC Name 1,2-bis(ethenyl)benzene;4-ethenylpyridine
Molecular Weight (g/mol) 235.33
PubChem CID 165743
Physical Form Powder
Infrared Spectrum Authentic
Packaging Plastic bottle
Water 10% max.
Color White to Cream
Quantity 50g
Chemical Name or Material Poly(4-vinylpyridine)
Show More Show Less