Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(acenaphthylene), ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
25036-01-5
MFCD00084391
25036-01-5
MFCD00084391
Physical Form Powder
Infrared Spectrum Authentic
Loss on Drying 2% max.
Packaging Glass bottle
Color Yellow
Quantity 5g
Chemical Name or Material Poly(acenaphthylene)