Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(acrylamide), carboxyl modified, pure, high carboxyl content, ∽ MW 200,000, ACROS Organics™

Click to view available options
Quantity:
5g
250g
Packaging:
Glass bottle
62649-23-4
C9H12NNaO5
237.187
MFCD00084393
DTBMFUOWAASNDB-UHFFFAOYSA-M
2-propenamide, polymer with 2-propenoic acid and sodium 2-propenoate,sodium acrylamide acrylic acid acrylate
23696299
sodium;prop-2-enamide;prop-2-enoate;prop-2-enoic acid
C=CC(=O)N.C=CC(=O)O.C=CC(=O)[O-].[Na+]
62649-23-4
C9H12NNaO5
237.187
MFCD00084393
DTBMFUOWAASNDB-UHFFFAOYSA-M
2-propenamide, polymer with 2-propenoic acid and sodium 2-propenoate,sodium acrylamide acrylic acid acrylate
23696299
sodium;prop-2-enamide;prop-2-enoate;prop-2-enoic acid
C=CC(=O)N.C=CC(=O)O.C=CC(=O)[O-].[Na+]
Physical Form Granules
Infrared Spectrum Authentic
Grade Pure
Loss on Drying 10% max.
Packaging Glass bottle
Solubility Solubility in water: soluble. Other solubilities: soluble in ethylene glycol, formamide, glycerol
Color White
Quantity 5g
Chemical Name or Material Poly(acrylamide), carboxyl modified