Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(acrylamide), granular, non-ionic, ∽ M.W. 5 to 6.000.000, ACROS Organics™

Click to view available options
Quantity:
5g
250g
500g
Packaging:
Glass bottle
9003-05-8,64-17-5
C3H5NO
71.08
MFCD00084392
HRPVXLWXLXDGHG-UHFFFAOYSA-N
acrylamide,2-propenamide,propenamide,acrylic amide,akrylamid,ethylenecarboxamide,vinyl amide,acrylic acid amide,2-propeneamide,propeneamide
6579
CHEBI:28619
prop-2-enamide
C=CC(=O)N
9003-05-8,64-17-5
C3H5NO
71.08
MFCD00084392
HRPVXLWXLXDGHG-UHFFFAOYSA-N
acrylamide,2-propenamide,propenamide,acrylic amide,akrylamid,ethylenecarboxamide,vinyl amide,acrylic acid amide,2-propeneamide,propeneamide
6579
CHEBI:28619
prop-2-enamide
C=CC(=O)N
Density 1.3000g/mL
Linear Formula [-CH2CH(CONH2)-]n
Formula Weight 5 to 6,000,000 (Average )
Physical Form Granules
Infrared Spectrum Authentic
Loss on Drying 13% max.
Packaging Glass bottle
Solubility Solubility in water: soluble. Other solubilities: soluble in ethylene glycol, glycerol, lactic acid
Specific Gravity 1.3
Color White to Yellow
Quantity 5g
Chemical Name or Material Poly(acrylamide)
Show More Show Less

RTECSNumber : AS3700000

TSCA : TSCA