Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(acrylic acid), 63 wt.% solution in water; approx. M.W. 2,000, ACROS Organics™

Click to view available options
Quantity:
5g
250g
500g
Packaging:
Glass bottle
9003-01-4,79-10-7,7732-18-5
C3H3NaO2
94.05
MFCD00084394
NNMHYFLPFNGQFZ-UHFFFAOYSA-M
acrylic acid,2-propenoic acid,propenoic acid,vinylformic acid,acroleic acid,propene acid,ethylenecarboxylic acid,polyacrylate,propenoate,carbomer
6581
CHEBI:18308
prop-2-enoic acid
C=CC(=O)O
9003-01-4,79-10-7,7732-18-5
C3H3NaO2
94.05
MFCD00084394
NNMHYFLPFNGQFZ-UHFFFAOYSA-M
acrylic acid,2-propenoic acid,propenoic acid,vinylformic acid,acroleic acid,propene acid,ethylenecarboxylic acid,polyacrylate,propenoate,carbomer
6581
CHEBI:18308
prop-2-enoic acid
C=CC(=O)O
Density 1.2300g/mL
Linear Formula [-CH2CH(CO2H)-]n
Physical Form Liquid
Infrared Spectrum Authentic
Percent Purity 60.0 to 65.0% (solids)
Packaging Glass bottle
Solubility Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, dioxane
Specific Gravity 1.23
Boiling Point 100.0°C
Color Undesignated
Quantity 250g
Chemical Name or Material Poly(acrylic acid), 63 wt. % Aqueous Solution
Show More Show Less

RTECSNumber : AT4680000

TSCA : TSCA